Anti-fkpA Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A51234-100UG A51234-50UG
77816-788EA 521.4 CAD
77816-788 77816-789
Anti-fkpA Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: fkpA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Escherichia coli O6:H1
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR