Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A49163-100UL A49163-50UL
77813-070EA 521.4 CAD
77813-070 77813-071
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, IHC: 1:50 to 1:200

Type:
Antigen: CXCR4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol CXCR4
Antigen name Fusion protein of CXCR4
Environmentally Preferable
Antigen synonyms C-X-C chemokine receptor type 4|CD184|CD184 antigen|Chemokine (C X C motif) receptor 4|Chemokine CXC Motif Receptor 4|CXC-R4|CXCR-4|CXCR4|CXCR4_HUMAN|D2S201E|FB22|Fusin|HM89|HSY3RR|LAP 3|LAP3|LCR1|LESTR|Leukocyte derived seven transmembrane domain receptor|Leukocyte-derived seven transmembrane domain receptor|Lipopolysaccharide associated protein 3|Neuropeptide Y receptor Y3|NPY3R|NPYR|NPYRL|NPYY3|NPYY3R|Probable G protein coupled receptor lcr1 homolog|SDF 1 receptor|SDF-1 receptor|SEVEN-TRANSMEMBRANE-SEGMENT RECEPTOR|Stromal cell derived factor 1 receptor|Stromal cell-derived factor 1 receptor|WHIM|WHIMS
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID P61073
Isotype IgG
ELISA Yes
ImmunoChemistry Yes
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 0.05% sodium azide, 40% glycerol, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR