Anti-UCP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: AFG Bioscience
A37990-100UG A37990-50UG
77811-937EA 521.4 CAD
77811-937 77803-937
Anti-UCP1 Rabbit Recombinant Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Type:
Antigen: UCP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol UCP1
Environmentally Preferable
Antigen name Mitochondrial brown fat uncoupling protein 1 protein (2-307AA)
Antigen synonyms mitochondrial brown fat uncoupling protein|Mitochondrial brown fat uncoupling protein 1|SLC25A7|Solute carrier family 25 member 7|Thermogenin|UCP 1|UCP|UCP1|UCP1_HUMAN|uncoupling protein 1 (mitochondrial, proton carrier)|Uncoupling protein 1
Clonality Recombinant Polyclonal
Conjugation FITC (Fluorescein Isothiocyanate)
Reactivity Human
Host Rabbit
Gene ID P25874
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at −20 or −80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR