Anti-UBD Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A37847-100UL
77811-823EA 924.3 CAD
77811-823
Anti-UBD Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: UBD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR