Anti-ZFP36 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A38669-100UL
77804-924EA 924.3 CAD
77804-924
Anti-ZFP36 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: ZFP36
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR