Anti-SDC1 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A34289-100UG A34289-50UG
77798-898EA 521.4 CAD
77798-898 77798-899
Anti-SDC1 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SDC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR