Anti-POLR3K Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31634-100UG A31634-50UG
77794-797EA 521.4 CAD
77794-797 77794-798
Anti-POLR3K Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: POLR3K
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol POLR3K
Antigen name DNA-directed RNA polymerase III subunit RPC10 protein (1-108AA)
Environmentally Preferable
Antigen synonyms C11|C11 RNP3|DNA directed RNA polymerase III subunit K|DNA directed RNA polymerase III subunit RPC10|DNA directed RNA polymerases III 12.5 kDa polypeptide|DNA-directed RNA polymerase III subunit K|DNA-directed RNA polymerase III subunit RPC10|hRPC11|HsC11p|My010|POLR3K|Polymerase (RNA) III (DNA directed) polypeptide K|12.3 kDa|RNA polymerase III 12.5 kDa subunit|RNA polymerase III subunit (hRPC11)|RNA polymerase III subunit C10|RNA polymerase III subunit C11|RNA polymerase III subunit CII|RPC10|RPC10_HUMAN|RPC11|RPC12.5
Clonality Recombinant Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID Q9Y2Y1
Isotype IgG
ELISA Yes
Form Liquid
Purification >95%, protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4.
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR