Saturix 3400SE Polyester Knit Dry Cleanroom Wipes, FG Cleanwipes
Supplier: FG CLEAN WIPES TE
7-3400SE-99L-S00 7-3400SE-BBL-S00
77786-132CS 643.06 CAD
77786-132 77786-134
Saturix 3400SE Polyester Knit Dry Cleanroom Wipes, FG Cleanwipes
Wipes Cleanroom Wipes Dry Cleanroom Wipes
FG Cleanroom Saturix 3400SE wipes are 100% polyester with sealed edges for minimal particle release. Designed for critical environments, they provide exceptional cleanliness, durability, and absorbency, ensuring effective contamination control in cleanrooms, labs, and precision manufacturing applications.
  • 100% polyester knit
  • Sealed edges
  • Heavyweight construction, 145 gsm
  • Double bagged in heavy mil, poly cleanroom packaging
  • Processed and packed in an ISO 4 cleanroom
  • High durability and chemical resistance
  • Economy and performance
  • Low NVRS, fibers, ions, and
More Product Information