Proteometer-L Starter Kit
Supplier: NOVILYTIC, LLC
KIT-PL-002-01
77775-510KT 5372 CAD
77775-510
Proteometer-L Starter Kit
Chromatography Columns
200 Titer + Aggregate assays. Use with any LC. For human mAbs, multi-specifics, recombinant proteins.
  • One assay for two critical attributes - titer + aggregate measurement
  • No protein A purification required: Reduces costs
  • No sample pretreatment: Reduces hours of sample prep
  • No method development: Reduces time until method usage
  • Direct analysis from CFB: HCP removal not needed
  • Plug-and-play for any LC with fluorescence detector: Easy adoption, no software needed
  • Low sample volume: Reduces impact on processing
Eliminate sample prep and method
More Product Information