Brady® i75 R6400 Ribbon for the i7500 Label Printer
Supplier: Brady Worldwide
176763 176764 176765 176766
77775-044RL 0 CAD
77775-044 77775-046 77775-048 77775-050
Brady® i75 R6400 Ribbon for the i7500 Label Printer
Printer Ribbons
i75-Series ribbon with LabelSense™ Technology warns user if ribbon is incompatible with installed i75-Series label material. Automatically prints at pre-tested heat and speed, ensuring correct print quality on the first label with no setting adjustments or trial-and-error print testing.
  • Ribbon in cartridge format
  • Ribbin ink wind direction: ink side out
  • Polyester material
  • Halogen-free
i75-series ribbon is self-centering with ink-side always correctly facing the label. One-color ribbon for printing a single color onto a label. R6400 Series high-density
More Product Information