Abgene™ Sample Processing and Storage Plates
Supplier: Thermo Fisher Scientific
AB-1178 AB-0765 AB-0781 AB-0787 AB-0788 AB-0796 AB-0859 AB-0932 AB-0988 AB-1056 AB-1058 AB-1127 AB-0564 AB-0661
77753-650CS 575.06 CAD
77753-650 77753-702 77753-686 77753-708 77753-674 77753-660 77753-652 77753-676 77753-710 77753-654 77753-682 77753-680 77753-698 77753-684
Abgene™ Sample Processing and Storage Plates
Microplates HTS Microplates
Abgene™ polypropylene MicroWell and DeepWell plates provide performance and value for sample storage, high-throughput screening, and molecular diagnostic assays. Abgene™ plates are made with high-quality, medical-grade, virgin polypropylene resin. These plates benefit from exceptional design and manufacturing advancements that ensure sample integrity and facilitate workflow efficiency.
  • Made of virgin, medical-grade polypropylene
  • hDNA-free, DNase-/RNase-free, and PCR inhibitor-free
  • Low extractables and leachables
  • 48 to 384 well plate formats available
More Product Information