Abgene™ Seals and Sealing Tape
Supplier: Thermo Fisher Scientific
AB-1720 AB-1797 AB-3559 AB-3720 AB-3738 AB-3739 AB-3797 AB-3799 AB-0745 AB-0757 AB-0812 AB-0559 AB-0580 AB-0718
77753-634CS 232.48 CAD
77753-634 77753-700 77753-638 77753-632 77753-704 77753-672 77753-688 77753-678 77753-662 77753-694 77753-640 77753-696 77753-664 77753-636
Abgene™ Seals and Sealing Tape
Seals Microplate Seals
Thermo Scientific™ Abgene™ seals minimize evaporation and protect samples from contamination and spilling. Adhesive seals seal all microplate formats and are ideal for all assay applications. Heat seals for ALPS™ microplate heat sealing instruments are ideal for storage applications, compound management, and high-throughput screenings including PCR.
  • Adhesive and heat seals
  • Temperature range −80 to +120 °C
  • Clear, white, and silver
  • Pierceable options
  • Peelable options
  • Gas permeable options
  • Heat seals designed for ALPS™ microplate heat sealers and generally
More Product Information