Human Recombinant FGF-22 (from E. coli)
Supplier: VWR International
77751-658 77751-660 77751-662 77751-664 77751-666
77751-658EA 1400.32 CAD
77751-658 77751-660 77751-662 77751-664 77751-666
Human Recombinant FGF-22 (from E. coli)
Proteins and Peptides
Fibroblast growth factor 22 (FGF-22) is a mediator of synaptogenesis in the adult nervous system and functions to regulate synapse formation and maturation. FGF-22 is expressed in the inner hair cell and functions to maintain ribbon synapse number to protect functional hearing. In the hippocampus, FGF-22 promotes excitatory synapse formation through binding the FGFR2b and FGFR1b receptors. FGF-22 is also required for axonal circuit remodeling after spinal cord injury.
  • High quality
  • Low endotoxin
  • Affordable
Caution: For research use only.