Human Recombinant PTN (from E. coli)
Supplier: VWR International
77751-592 77751-594 77751-596 77751-598 77751-600
77751-592EA 949.5 CAD
77751-592 77751-594 77751-596 77751-598 77751-600
Human Recombinant PTN (from E. coli)
Proteins and Peptides
Pleiotrophin (PTN) is a heparin-binding growth factor that has mitogenic effects on fibroblast, epithelial, and endothelial cells. PTN is made by many tissues, but is predominantly secreted by nervous tissue during development. PTN induces neurite outgrowth and is involved in tumor growth and metastasis. PTN binds with low affinity to the cell surface receptor nucleolin to inhibit HIV-1 infection. PNT also binds the receptor protein tyrosine phosphatase type Z (PTPRZ), syndecan-3, and anaplastic lymphoma kinase (ALK) receptors.
  • High quality
  • Low endotoxin
  • Affordable
Caution:
For research use only.