Anti-HCFC2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24431-100UL
77739-495EA 924.3 CAD
77739-495
Anti-HCFC2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: HCFC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR