Anti-MCL1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A24305-50UG
77739-380EA 355.5 CAD
77739-380
Anti-MCL1 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500 to 1:2000, ELISA:1:10000

Type:
Antigen: IER2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR