Anti-SLC4A9 Rabbit Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A63832-50UG A63832-100UG
77722-887EA 360.84 CAD
77722-887 77722-886
Anti-SLC4A9 Rabbit Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: SLC4A9
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol SLC4A9
Environmentally Preferable
Antigen synonyms SLC4A9 solute carrier family 4|sodium bicarbonate cotransporter like|Anion exchanger 4|B3A4_HUMAN|Sodium bicarbonate cotransporter 5|Solute carrier family 4 member 9|Anion exchange protein 4|AE4|Slc4a9|SBC5|solute carrier family 4|AE 4|member 9|sodium bicarbonate cotransporter
Clonality Polyclonal
Conjugation Biotin
Reactivity Human
Host Rabbit
Gene ID Q96Q91
Isotype IgG
ELISA Yes
Immunogen Recombinant Human Anion exchange protein 4 protein (114-402AA)
Form Liquid
Purification >95%, Protein G purified
Storage buffer 0.03% Proclin 300, 50% glycerol, 0.01 M PBS, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on Blue Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR