Anti-SYNE2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A63545-100UG A63545-50UG
77722-337EA 529.22 CAD
77722-337 77722-338
Anti-SYNE2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SYNE2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR