Anti-BLNK Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A63503-100UG A63503-50UG
77722-258EA 529.22 CAD
77722-258 77722-259
Anti-BLNK Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: BLNK
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR