Anti-CSF1R Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A20069-100UG
77701-922EA 400.93 CAD
77701-922
Anti-CSF1R Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, ELISA:1:10000

Type:
Antigen: CSF1R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
New Product
Order Now

SPECIFICATIONS

Antigen symbol CSF1R
Environmentally Preferable
Antigen synonyms MCSFR|Oncogen FMS|CD115|CSF-1-R|CSF 1 R|CSF-1 receptor|CSF1 R|CD115 antigen|FIM 2|FMS protooncogene|CSFR|EC 2.7.10.1|CFMS|HDLS|CSF 1R|V-FMS McDonough feline sarcoma viral oncogen homolog|CSF1R_HUMAN|C FMS|CD 115|FMS proto oncogene|Macrophage colony stimulating factor I receptor|Colony stimulating factor 1 receptor|Colony stimulating factor I receptor|M-CSF Receptor|Proto-oncogene c-Fms|Macrophage colony-stimulating factor 1 receptor|McDonough feline sarcoma viral (v fms) oncogene homolog|M-CSF-R|FMS|Macrophage colony stimulating factor 1 receptor|FIM2|CSF1R|formerly
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID P07333
Isotype IgG
Western blot Yes
ELISA Yes
Immunogen Synthesized peptide derived from Human c-Fms around the non-phosphorylation site of Y699.
Form Liquid
Purification Antigen affinity purified
Storage buffer PBS, 50% glycerol, 0.5% BSA, 0.02% sodium azide, pH 7.4
Storage temperature Upon receipt, store at –20 or –80 °C. Avoid repeated freeze.
Shipping temperature Shipped on blue ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR