Anti-MERTK/TYRO3 (phospho-Y749/681) Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A19012-100UG
77701-560EA 400.93 CAD
77701-560
Anti-MERTK/TYRO3 (phospho-Y749/681) Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000

Type:
Antigen: MERTK/TYRO3 (phospho-Y749/681)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR