Anti-NDUFB9 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A14752-100UG
77700-062EA 400.93 CAD
77700-062
Anti-NDUFB9 Rabbit Polyclonal Antibody
Antibodies
WB:1:500-1:2000, IHC:1:100-1:300, ELISA:1:10000

Type:
Antigen: NDUFB9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR