Anti-YIPF5 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A63628-100UG A63628-50UG
77690-298EA 529.22 CAD
77690-298 77722-492
Anti-YIPF5 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:1000-1:5000, IF:1:50-1:200

Type:
Antigen: YIPF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR