Anti-pvdA Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A57153-100UG
77689-734EA 529.22 CAD
77689-734
Anti-pvdA Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: WB:1:500-1:5000

Type:
Antigen: pvdA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pseudomonas aeruginosa
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR