Anti-TPPP2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A37317-100UG
77688-824EA 529.22 CAD
77688-824
Anti-TPPP2 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: TPPP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR