Starter Pen Meters for Conductivity
Supplier: Ohaus
30073974 30073970 30073971 30073972 30073973 30073975 30073976 30073977
77679-532EA 168.36 CAD
77679-532 77681-980 77681-982 77681-984 77681-986 77681-988 77681-990 77681-992
Starter Pen Meters for Conductivity
Electrochemistry Meters Conductivity Meters
Starter pen meters are the economical option when you are simply looking for meters that will provide accurate measurement without restriction.
  • Easily replaceable electrodes
  • Automatic shut off feature
  • Liquid crystal display (LCD) display with secondary information line (on select models)
OHAUS precision is now available in a line of pen meters that provide accurate measurement of pH, oxidation-reduction potential (ORP), conductivity, salinity, and total dissolved solids (TDS).