PUREZERO* HG3 Cream Latex Gloves
Supplier: O&M HALYARD CANADA, INC CA
CLN823280 CLN823285 CLN823290 CLN8231LG CLN8231MD CLN8231SM CLN8231XL CLN8231XS CLN823210 CLN823260 CLN823265 CLN823270 CLN823275
77681-966CS 539.3 CAD
77681-966 77681-968 77681-970 77681-946 77681-948 77681-950 77681-952 77681-954 77681-956 77681-958 77681-960 77681-962 77681-964
PUREZERO* HG3 Cream Latex Gloves
Gloves
PUREZERO* HG3 cream latex gloves are designed specifically to meet the needs of semi-conductor electronics medical device and other cleanroom manufacturing applications.
  • Recommended for use in ISO Class 3 or higher and Grade B/C/D cleanrooms
  • Maximum Particles 0.5 µm/cm² less than 1200
  • 30.2 cm (12") length with beaded cuff available in hand-specific (sterile) and ambidextrous (non-sterile) options
  • AQL 1.5 for pinholes
  • Silicone and powder free
  • Tested against 35 chemicals 14 chemotherapy drugs and fentanyl
  • Recommended for applications such as semi-conductor
More Product Information