AquaSearcher™ AP30TUR Portable Turbidimeters
Supplier: Ohaus
30853478 30853396 30853397 30853395
77679-514EA 1491.63 CAD
77679-514 77679-510 77679-512 77679-508
AquaSearcher™ AP30TUR Portable Turbidimeters
Turbidity Meters
AquaSearcher AP30TUR series portable turbidimeter combines two methods, eliminating the need to buy two different meters for your water measurement applications. This meter is equipped with dual optic paths and detectors to ensure accuracy to comply with industry standards. AP30TUR is built robustly for field applications.
  • Combines the nephelometric and transmitted method
  • Water-resistant, has a long battery life and can store up to 5000 measurements
  • Users can easily transfer data to a PC with USB connection without requiring additional software
  • Built-in
More Product Information