ECO Containers for ECO Funnels, CP Lab Safety
Supplier: CP Lab Safety
EC-1275120 EC-1275450 EC-2002 EC-2009 EC-20010 EC-20020 EC-20020DRN EC-20020DRB EC-38400 EC-384004 EC-38GL-2 EC-38GLSC EC-63RC
33500-978EA 817.35 CAD
33500-978 33500-982 33501-868 33501-874 33501-876 33501-878 33501-880 33501-882 33501-884 77676-674 77676-676 77676-678 77676-682 77676-684 77676-686
ECO Containers for ECO Funnels, CP Lab Safety
Sample Containers General Purpose Containers
ECO Funnels® are designed to fit a wide variety of standard Nalgene® carboys, Justrite® poly waste cans and other containers. The selection below includes the most common containers used with the ECO Funnel laboratory.
  • Made from PE
  • Good chemical resistance when glass is not an option
Containers with 38 mm, 53 mm, and 70 mm openings will typically work with one of the ECO Funnels.
Order Now
SPECIFICATIONS
Environmentally Preferable