Clariflow® B Mini-Capsule Filters, Parker Hannifin
Supplier: PARKER HANNIFIN TE
25-CDBBB-002-2 25-CDBBB-002-2-S 25-CDBHH-002-2-S 25-CDBSS-002-2 25-CDBSS-002-2-S 25-CHBBB-002-2 25-CHBBB-002-2-S 25-CHBHH-002-2 25-CHBHH-002-2-S 25-CHBSS-002-2 25-CHBSS-002-2-S 25-CSBBB-002-2 25-CSBBB-002-2-S 25-CSBHH-002-2 25-CSBHH-002-2-S 25-CSBSS-002-2 25-CSBSS-002-2-S
77676-222EA 408.21 CAD
77676-222 77676-226 77676-236 77676-238 77676-246 77676-230 77676-220 77676-242 77676-244 77676-218 77676-228 77676-248 77676-234 77676-240 77676-216 77676-232 77676-224
Clariflow® B Mini-Capsule Filters, Parker Hannifin
Filters Membrane Filter Capsules
Clariflow-B Mini-Capsules deliver high-purity fluids for small volume biopharmaceutical applications. The mirrored anisotropic polyethersulfone (PES) membrane is biologically inert low protein-binding and hydrophilic.
  • High flow rate reduces processing time
  • Biologically inert, low protein binding membrane maximizes product yield
  • Application includes: sera and vaccines, bacterial growth media, parenteral solutions, buffers and reagents, protein-containing preparartions
Clariflow-B Mini-Capsules are available in a variety of sizes and end-fittingMore Product Information