CHEMets® Ammonia Visual HR Test Kit (Nessler 1000 to 10000 ppm)
Supplier: CHEMetrics
K-1520C
77674-728EA 145.36 CAD
77674-728
CHEMets® Ammonia Visual HR Test Kit (Nessler 1000 to 10000 ppm)
Environmental Test Kits
Delivery information: The K-1520C ammonia HR CHEMets® test kit (1000 to 10,000 ppm) comes in a plastic case and contains everything needed to perform 30 tests (except distilled water): 30 HR CHEMets® ampoules, a color comparator, fixed volume pipet, 25 ml sample cup, and instructions. This kit is applicable for wastewater, environmental, aquaculture or any application where high concentrations of ammonia may be found in water.
Caution: Contains mercury. Dispose according to local, state or federal laws.