Sterilisation Baskets, Side Perforated, Flat Base
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP-4553 WP-4554 WP-4555 WP-4556 WP-4557 WP-4558 WP-4559 WP-4560 WP-4561 WP-4562 WP-4563 WP-4564 WP-4565 WP-4566 WP-4567 WP-4568 WP-4569 WP-4570 WP-4571 WP-4572 WP-4545 WP-4546 WP-4547 WP-4548 WP-4549 WP-4550 WP-4551 WP-4552
77666-156EA 161.16 CAD
77666-156 77666-158 77666-160 77666-162 77666-164 77666-166 77666-168 77666-170 77666-172 77666-174 77666-176 77666-178 77666-180 77666-182 77666-184 77666-186 77666-188 77666-190 77666-192 77666-194 77666-140 77666-142 77666-144 77666-146 77666-148 77666-150 77666-152 77666-154
Sterilisation Baskets, Side Perforated, Flat Base
Baskets Wire Baskets
Stainless steel sterilization baskets for instrument cleaning, sterilization and storage. The perforated side walls protect instrument edges and tips from damage. These sterilization baskets feature electropolished smooth, rounded corners. The baskets are burr free to protect wraps and prevent injury.