Full Micro Perforated Trays with Lids
Supplier: WORLD PRECISION INSTRUMENTS LLC
WP-2343 WP-2354 WP-2355 WP-2356 WP-2357 WP-2358 WP-2359 WP-2344 WP-2345 WP-2346 WP-2347 WP-2348 WP-2349 WP-2350 WP-2351 WP-2352 WP-2353
77665-634EA 53.72 CAD
77665-634 77665-656 77665-658 77665-660 77665-662 77665-664 77665-666 77665-636 77665-638 77665-640 77665-642 77665-644 77665-646 77665-648 77665-650 77665-652 77665-654
Full Micro Perforated Trays with Lids
Trays/Dishes/Pots Instrument Trays/Dishes
Full micro perforated trays with lids manufactured of stainless steel grade AISI-30.
  • Electropolished
  • Full wall perforation to protect sharp instruments
  • Protects edges and tips from damage
  • Smooth rounded corners
  • Perforation hole size is 2.0 mm
  • Hinged lids
Top and lid frame rod diameter: 3.5 mm and 4.0 mm (depending on the basket). Full wall perforation to protect sharp instrument's edges and tips from damage. Fine mesh with opening squares of 1.6 mm and wire diameter of 0.7 mm. Sheet locking clip.
Applications:
Micro surgical instrument cleaning andMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable