Microinjection Syringe Pumps, UMP3
Supplier: WORLD PRECISION INSTRUMENTS LLC
UMP3 UMP3-3 UMP3-4
77665-468EA 5734.83 CAD
77665-468 77665-470 77665-472
Microinjection Syringe Pumps, UMP3
Pumps Syringe Pumps
Versatile UMP3 micro syringe pump injector that uses micro syringes to deliver microliter and nanoliter volumes.
  • Syringes are easily installed, just snap the barrel into the clamps
  • Threaded shaft drives syringe plunger allowing smooth and accurate movement
  • Total # of steps: 20,000 (63 mm travel)
  • Minimum dispensing volume: 25 nl
  • Linear motion per step: 3.175 μm
With its digital controller, UltraMicroPump III can dispense as little as 600 picoliters per incremental advance of the syringe piston (using a 0.5 μl syringe). Syringes may be
More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable