Accupulser™ A310 Signal Generator
Supplier: WORLD PRECISION INSTRUMENTS LLC
SYS-A310
77665-170EA 11267.38 CAD
77665-170
Accupulser™ A310 Signal Generator
Batteries
The A310 pulse generator/stimulator combines the reproducibility and accuracy of digital electronics with the fine resolution and continuous adjustment pos­si­ble with analog circuitry. All timing parameters are entered with high resolution, ten-turn potentiometers and six-position range switches. Timing is ac­cu­rate to within 1% of the set value.
  • Single channel pulse generator with train capability
  • TTL and variable voltage output
  • Variety of pulses: continuous run, single-shot, train/burst
  • Multiple outputs available: monitor, isolator, sync and variable
More Product Information