Rodent Rectal Temperature Probes
Supplier: WORLD PRECISION INSTRUMENTS LLC
RET-2 RET-3
77665-122EA 559.95 CAD
77665-122 77665-124
Rodent Rectal Temperature Probes
Temperature Probes
Flexible Teflon microprobes are used for implantation in tissue, in spectrophotometer cuvettes, rectally in neonatal mice, in water baths, PCR thermal cyclers, etc. Also, measure animal rectal temperatures during surgical procedures and pyrogen testing or skin temperature measurement during exercise physiology studies.
  • Works with the ATC2000 TC input
  • Not isolated
  • 125 °C Max. Temperature
  • 5 ft. lead length
When precise temperature measurements are required, WPI can provide you with a very accurate monitor and thermocouple microprobes. WPI monitors haveMore Product Information