Microelectrode Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
MPH110 MPH112 MPH115 MPH120 MPH310 MPH312 MPH315 MPH320 MPH410 MPH412 MPH415 MPH420
77664-838EA 284.4 CAD
77664-838 77664-840 77664-842 77664-844 77664-846 77664-848 77664-850 77664-852 77664-854 77664-856 77664-858 77664-860
Microelectrode Holders
Electrode Holders
WPI™s microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers.
  • Male connector
  • Pellet half-cell
  • No pressure port
  • Screw cap
Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps).
Filling WPI microelectrode holders with electrolytes containing chloride resultsMore Product Information