Microelectrode Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
MEH3SFW10 MEH3SFW12 MEH3SFW15 MEH3SFW20
77664-720EA 199.54 CAD
77664-720 77664-722 77664-724 77664-726
Microelectrode Holders
Electrode Holders
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets.
  • Female connector
  • Wire half-cell
  • No pressure port
  • Screw cap
The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubber gaskets (without caps). Filling WPI microelectrode holders with electrolytes containing chloride resultsMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable