Microelectrode Holders
Supplier: WORLD PRECISION INSTRUMENTS LLC
MEH2RW10 MEH2RW12 MEH2RW15
77664-638EA 319.79 CAD
77664-638 77664-640 77664-642
Microelectrode Holders
Electrode Holders
All holders are supplied for standard WPI single capillary tubing of 1.0, 1.2, 1.5 and 2.0 mm outside diameters.
  • Male connector
  • Wire half-cell
  • Male luer
  • Screw cap
WPI's microelectrode holder-half-cells couple fluid-filled glass micropipettes to high input impedance amplifiers. A Ag/AgCl pellet (or a silver wire) molded into the holder body provides stable potential. Electrical connection is made via male 2 mm pins or female 2 mm sockets. The pipette may be mounted axially or at right angles to the holder. Pipettes are held with screw-caps or rubberMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable