Manual Micromanipulator, M3301
Supplier: WORLD PRECISION INSTRUMENTS LLC
M3301L M3301-M3-L M3301R
77664-578EA 3990.94 CAD
77664-578 77664-580 77664-584
Manual Micromanipulator, M3301
Micromanipulators
The M3301 is a popular Manual Micromanipulator, because it is accurate, lightweight and well-engineered. This solid manipulator outsells all others worldwide for high precision experiments where magnification is in the range of up to 250×.
  • Lightweight (550 g)
  • Sure, repeatable movement without drift
  • Choice of optional M-3 tilting base
  • Thumbscrew included for metric anti-vibration platforms with M6 holes
  • 5464 Base Weight and 15873 Angled Electrode Holder sold separately
  • Comes installed with a 12 mm clamp for the M10 magnetic base
It has a slim, space-More Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable