Dri-Ref™ Reference Electrodes
Supplier: WORLD PRECISION INSTRUMENTS LLC
SDR2 DRIREF-2 DRIREF-2SH DRIREF-450 DRIREF-5 DRIREF-5SH DRIREF-L FLEXREF
77665-138EA 346.02 CAD
77665-138 77664-216 77664-218 77664-220 77664-222 77664-224 77664-226 77664-338
Dri-Ref™ Reference Electrodes
Electrodes ISE Electrodes
Dri-Ref™ reference electrodes have extremely low electrolyte leak­age properties and exhibit stable and reproducible potential and low resistance.
  • Stored in KCl when not in use, they have a long life expectancy
  • Extremely low electrolyte leakage
  • Stable, reproducible potential with low resistance
  • Chemically resistant to strong acids and bases
  • Ideal for small volume, low salt concentration measurement (SUPER-Dri-Ref)
May be used with ion selective electrodes without contamination from the reference electrode. Although the internal filling solutionMore Product Information