Rodent Guillotines
Supplier: WORLD PRECISION INSTRUMENTS LLC
DCAP-M DCAP DCAP-L
77664-196EA 1963.94 CAD
77664-196 77664-192 77664-194
Rodent Guillotines
Knives
These guillotines are designed for small animal researchers, and they come in three sizes. The guillotine is considered one of the most humane methods of dispatching laboratory animals. With a single downward motion of the handle, you can euthanize a rodent quickly and without trauma.
  • Guillotine designed for small animal research
  • Three sizes for mice, rats and other small animals
  • Large, stable base
  • Hardened blades ensure you get many years of service and the blades remain sharp
  • Ambidextrous configuration allows comfortable use for right and left handed
More Product Information