Modular Transducer Amplifier, Bridge8
Supplier: WORLD PRECISION INSTRUMENTS LLC
BRIDGE8
77664-076EA 2607 CAD
77664-076
Modular Transducer Amplifier, Bridge8
Data Loggers
Bridge8 is a modular, rack-mountable amplifier system. It is specifically designed for use as a signal conditioning amplifier with strain gages and other poered transducers. It includes differential amplifiers featuring high input impedance, high common mode rejection and low current leakage input terminals for low noise operation.
  • Wide range of fixed gains with independent variable gain adjustment
  • Low pass filter
  • Single ended or differential transducer compatibility
  • Dual range output offset correction
  • Independent module power switch
  • Provides ± voltage
More Product Information