Six Programble Pump, Aladdin AL-6000
Supplier: WORLD PRECISION INSTRUMENTS LLC
AL-6000
77664-056EA 4911.11 CAD
77664-056
Six Programble Pump, Aladdin AL-6000
Pumps Syringe Pumps
Aladdin AL-6000 holds up to six syringes. While it is one of the most cost-effective pumps available, the Aladdin pump series boasts versatility and functionality to serve a variety of dispensing applications!
  • Cost-effective
  • Program sequences without a computer
  • Rich command set
  • RS-232 and TTL control
  • Accepts syringe sizes: 1 to 60 ml, or 0.5 to 5 ul
  • Multi-Barrel pump for up to six syringes
Benefits:
Automatic dispensing of small volumes
Very precise flow rate control
Hands-free operation with foot switch
Applications:
Drug administration
ChemicalMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable