AirTherm™ SMT Heat Controller/Humidity Monitors
Supplier: WORLD PRECISION INSTRUMENTS LLC
AIRTHERM-SMT-1W AIRTHERM-SMT-2W
77664-040EA 7892.1 CAD
77664-040 77664-042
AirTherm™ SMT Heat Controller/Humidity Monitors
Controllers Temperature Controllers
The AirTherm™ SMT is a new generation of heat control system from WPI designed to be used in Live Cell Imaging applications with microscopes fitted with a full microscope environmental chamber enclosure. The standard AirTherm™ SMT controls temperature and, as an option, monitors humidity.
  • Precision heat controller for use in live cell imaging and custom incubators
  • Control heat and monitor humidity (optional) with a single controller
  • Electrically and acoustically quiet
  • Quick, precise response to thermal change
Benefits:
PID control algorithms allow forMore Product Information