PTFE Coated Silver Wires
Supplier: WORLD PRECISION INSTRUMENTS LLC
AGT0510 AGT05100 AGT0525 AGT1010 AGT10100 AGT1025 AGT1510 AGT1530
77664-004EA 135.88 CAD
77664-004 77664-006 77664-008 77664-010 77664-012 77664-014 77664-016 77664-018
PTFE Coated Silver Wires
Wire
PTFE coated silver wire is available is precut lengths. It is design to reduce surface friction. Available in 36, 30, and 26 to 27 AWG.
The PTFE coating is 150 micro inches thickness (4 μm). It is designed to reduce surface friction, only. It is not insulation.
Annealed silver wire is 99.99% pure and and PTFE coated (coating adds 0.002 inches to diameter of wire).