Box Filaments
Supplier: WORLD PRECISION INSTRUMENTS LLC
14074 13834 506122 505095 505096 505097 505098
77661-700EA 271.82 CAD
77661-700 77661-648 77663-860 77663-820 77663-822 77663-824 77663-826
Box Filaments
Puller and Inserter Tools
A glass puller is a laboratory instrument used to create fine, tapered glass micropipettes or microelectrodes. The choice of filament for the glass puller is crucial, as it directly influences the heating element's performance and, consequently, the quality of the pulled glass. Different types and shapes of filaments can be used, and the selection depends on factors such as the desired pipette.
  • Filament, platinum/iridium
  • Variety of sizes
The box filament is configured in the shape of a square, allowing for controlled heating in the corners or alongMore Product Information
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable