Catherization Cannulation Kit
Supplier: WORLD PRECISION INSTRUMENTS LLC
504742
77663-758EA 934.27 CAD
77663-758
Catherization Cannulation Kit
Catheters
Nine piece surgical instrument kit ideal for catherization and cannulation.
Delivery information: Kit includes: Disposable scalpel, #15, sterile, 10/box, lever action skin stapler 35S, sterile, Metzenbaum scissors, 11.5 cm, SuperCut, straight, castroviejo needle holder with lock, 14 cm, straight, serrated, Halsted mosquito hemostatic forceps, 12.5 cm, straight, Spence stitch ligature scissors, 9 cm, Adson-Brown forceps, 12 cm, straight, TC jaws, 7×7 teeth, WPI Swiss tweezers #5, 11 cm, 0.1×0.06 mm tips, Nugent utility forceps, 10 mm, angled tip.
New Product
Order Now
SPECIFICATIONS
Environmentally Preferable