Katena-Vannas Spring Scissors, 11 cm
Supplier: WORLD PRECISION INSTRUMENTS LLC
504024 504025 504026 504027
77663-440EA 717.75 CAD
77663-440 77663-442 77663-444 77663-446
Katena-Vannas Spring Scissors, 11 cm
Scissors
Katena Vannas spring scissors are microsurgical spring scissors used in delicate ophthalmic procedures and other microsurgical applications in a research laboratory. They are named after the renowned Swiss ophthalmic surgeon, Nils Yngve Vannas.
  • Blades are thin, sharp and pointed, 7 or 10 mm long
  • Curved or straight tips
Spring scissors can be used ambidextrously, making them favored choice for left-handed people. These scissors are known for their high-quality construction and extreme precision, making them valuable tools in eye surgery.