Volkman Bone Curette
Supplier: WORLD PRECISION INSTRUMENTS LLC
503689
77663-360EA 92.46 CAD
77663-360
Volkman Bone Curette
Curettes
The Volkman bone curette is a bone curette for debulking, and it has a razor-sharp cutting edge. It is commonly used in orthopedic procedures.
  • Oval cutting cup (1 cm or 5 mm) or double ended curette
The knurled round handle of this curette gives you a sure grip. The handle gradually tapers to the tips. Removal of fibrous tissue from bone or scrape bone to remove small pieces or shape it with this curette. This cutting edge is an oval scoop at the end of the curette. This is manufactured for surgical grade stainless steel and is suitable forMore Product Information